Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%

Supplier: AMBEED, INC
Warning

A151105-1G A151105-5G A151105-10G A151105-250MG A151105-1G A151105-25G A151105-5G
103804-232EA 29 USD InStock
103804-232 103842-378 77010-760 77010-762 77317-918 77317-920 77317-922
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₇N₃O₂S
Boiling Pt: 412.8 °C (760 mm Hg)
Flash Pt: 203.4 °C
Storage Temperature: Ambient
MDL Number: MFCD08448157
CAS Number: 365996-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty