Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-aminobenzylcarbamate 95%

Supplier: AMBEED, INC
Warning

A172826-1G A172826-5G A172826-1G A172826-5G A172826-250MG
103806-752EA 114 USD InStock
103806-752 103845-044 77082-256 77082-258 77138-466
tert-Butyl 2-aminobenzylcarbamate 95%
tert-Butyl 2-aminobenzylcarbamate
Precaución: for Research Use Only


Formula: C₁₂H₁₈N₂O₂
MW: 222.28 g/mol
Boiling Pt: 382.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04071350
CAS Number: 162046-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty