Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine 95%

Supplier: AMBEED, INC
Warning

A173750-5G A173750-10G A173750-250MG A173750-1G A173750-25G A173750-5G
103806-866EA 18 USD InStock
103806-866 103845-166 76784-326 76784-328 77082-436 77082-438 77082-440
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine 95%
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine
Precaución: for Research Use Only


Formula: C₁₁H₁₅BrN₂O₂S
Melting Pt: 87…91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09951958
CAS Number: 365996-06-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty