Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-2-bromopropionate 98%

Supplier: AMBEED, INC
Warning

A121132-100G A121132-10G A121132-25G A121132-5G
77258-102EA 163 USD InStock
77258-102 77258-104 77258-106 77312-150
tert-Butyl-2-bromopropionate 98%
tert-Butyl-2-bromopropionate
Precaución: for Research Use Only


Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
Boiling Pt: 170.2±8.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00043897
CAS Number: 39149-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty