Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-cyanomorpholine-4-carboxylate 98%

Supplier: AMBEED, INC
Warning

A105802-1G A105802-5G A105802-100MG A105802-250MG A105802-1G
103796-948EA 158 USD InStock
103796-948 103822-692 76821-538 76821-540 77309-556
tert-Butyl 2-cyanomorpholine-4-carboxylate 98%
tert-Butyl 2-cyanomorpholine-4-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₆N₂O₃
Boiling Pt: 340.3±42.0 °C (760 mm Hg)
Flash Pt: 159.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18250325
CAS Number: 1211592-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty