Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-2-cyanopyrrolidine-1-carboxylate 95%

Supplier: AMBEED, INC
Warning

A366465-100G A366465-10G A366465-1G A366465-5G A366465-25G
103826-400EA 12 USD InStock
103826-400 103837-416 76817-444 76817-446 76840-650 76840-652 77306-774
tert-Butyl-2-cyanopyrrolidine-1-carboxylate 95%
tert-Butyl-2-cyanopyrrolidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₆N₂O₂
MW: 196.2500 g/mol
Boiling Pt: 307.9 °C (760 mm Hg)
Melting Pt: 33…36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861225
CAS Number: 144688-70-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty