Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-ethynylbenzylcarbamate 95%

Supplier: AMBEED, INC
Warning

A304022-1G A304022-100MG A304022-250MG A304022-1G
103814-408EA 581 USD InStock
103814-408 77014-996 77014-998 77305-892
tert-Butyl 2-ethynylbenzylcarbamate 95%
tert-Butyl 2-ethynylbenzylcarbamate
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₂
MW: 231.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24471043
CAS Number: 1097731-47-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty