Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A234519-1G A234519-5G A234519-1G A234519-25G A234519-5G A234519-100G A234519-10G
103810-956EA 26 USD InStock
103810-956 103850-712 77089-698 77089-700 77089-702 77098-314 77098-316
tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate 97%
tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂O₃
Boiling Pt: 322.8 °C (760 mm Hg)
Melting Pt: 92…96 °C
Flash Pt: 149.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04115304
CAS Number: 301673-16-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty