Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-bromo-6-chloropicolinate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A146851-1G A146851-5G A146851-250MG A146851-1G A146851-5G A146851-25G
103803-710EA 30 USD InStock
103803-710 103841-804 76823-674 77317-150 77317-152 77683-255
tert-Butyl 3-bromo-6-chloropicolinate 97%, Ambeed.Inc
tert-Butyl 3-bromo-6-chloropicolinate
Precaución: For research use only.


Formula: C₁₀H₁₁BrClNO₂
MW: 292.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19443279
CAS Number: 1235036-15-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty