Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-bromopicolinate 97%

Supplier: AMBEED, INC
Warning

A100365-1G A100365-5G A100365-250MG A100365-25G A100365-1G A100365-5G
103795-390EA 47 USD InStock
103795-390 103821-090 77135-434 77135-436 77282-482 77282-484
tert-Butyl 3-bromopicolinate 97%
tert-Butyl 3-bromopicolinate
Precaución: for Research Use Only


Formula: C₁₀H₁₂BrNO₂
MW: 258.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264589
CAS Number: 1260807-49-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty