Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-bromopropanoate 98%

Supplier: AMBEED, INC
Warning

A200266-1G A200266-5G A200266-100G A200266-10G A200266-250MG A200266-25G A200266-1G A200266-5G
103817-182EA 6 USD InStock
103817-182 103847-810 76814-264 76814-266 76814-268 76814-270 77085-870 77302-670
tert-Butyl 3-bromopropanoate 98%
tert-Butyl 3-bromopropanoate
Precaución: for Research Use Only


Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
Boiling Pt: 188.8 °C (760 mm Hg)
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974087
CAS Number: 55666-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty