Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-ethynylbenzoate 95%

Supplier: AMBEED, INC
Warning

A102187-1G A102187-5G A102187-100MG A102187-250MG A102187-1G A102187-5G
103796-328EA 87 USD InStock
103796-328 103822-022 77135-538 77135-540 77308-896 77308-898
tert-Butyl 3-ethynylbenzoate 95%
tert-Butyl 3-ethynylbenzoate
Precaución: for Research Use Only


Formula: C₁₃H₁₄O₂
MW: 202.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25542332
CAS Number: 914943-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty