Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-hydroxyphenethylcarbamate 97%

Supplier: AMBEED, INC
Warning

A101127-1G A101127-100MG A101127-250MG
103795-814EA 84 USD InStock
103795-814 77135-478 77135-480
tert-Butyl 3-hydroxyphenethylcarbamate 97%
tert-Butyl 3-hydroxyphenethylcarbamate
Precaución: for Research Use Only


Formula: C₁₃H₁₉NO₃
MW: 237.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24448669
CAS Number: 129150-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty