Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 98%

Supplier: AMBEED, INC
Warning

A245654-1G A245654-5G A245654-1000G A245654-100G A245654-10G A245654-250MG A245654-500G A245654-1G A245654-25G A245654-5G
103811-660EA 6 USD InStock
103811-660 103851-492 76722-530 76722-532 76722-534 76722-536 76722-538 77090-752 77090-754 77090-756
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 98%
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₉NO₃
Boiling Pt: 325.8 °C (760 mm Hg)
Melting Pt: 70…74 °C
Flash Pt: 150.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00673779
CAS Number: 185099-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty