Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine-1-carboxylate 96%

Supplier: AMBEED, INC
Warning

A753614-5G A753614-1G A753614-100MG A753614-250MG
103840-858EA 179 USD InStock
103840-858 76845-090 76981-376 76981-378
tert-Butyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine-1-carboxylate 96%
tert-Butyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine-1-carboxylate
Precaución: For research use only


Formula: C₂₁H₃₃BN₂O₄
Boiling Pt: 508.8±45.0 °C (760 mm Hg)
Flash Pt: 261.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781212
CAS Number: 540752-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty