Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-piperidin-4-ylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A218354-1G A218354-5G A218354-10G A218354-250MG A218354-1G A218354-5G
103818-708EA 9 USD InStock
103818-708 103849-582 76814-820 76814-822 77303-486 77303-488
1-Boc-piperidin-4-ylboronic acid pinacol ester 97%
1-Boc-piperidin-4-ylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₆H₃₀BNO₄
MW: 311.2247 g/mol
Boiling Pt: 358.8 °C (760 mm Hg)
Melting Pt: 60…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997792
CAS Number: 1048970-17-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty