Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A249692-1G A249692-10G A249692-250MG A249692-1G A249692-25G A249692-5G
103811-884EA 10 USD InStock
103811-884 103851-722 76987-852 76987-854 76987-856 77091-060 77091-062 77091-064
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate 97%
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₆H₂₄N₂O₂
MW: 276.3740 g/mol
Melting Pt: 187…188 °C
Storage Temperature: Ambient
MDL Number: MFCD05861405
CAS Number: 170011-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty