Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-ylpiperazine-1-carboxylic acid tert-butyl ester 97%

Supplier: AMBEED, INC
Warning

A173531-1G A173531-5G A173531-1G A173531-25G A173531-5G A173531-100G A173531-10G A173531-250MG
103806-824EA 10 USD InStock
103806-824 103845-124 77082-366 77082-368 77082-370 77163-642 77163-644 77163-646
4-[5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-ylpiperazine-1-carboxylic acid tert-butyl ester 97%
4-[5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-ylpiperazine-1-carboxylic acid tert-butyl ester
Precaución: for Research Use Only


Formula: C₂₀H₃₂BN₃O₄
Boiling Pt: 515.9 °C (760 mm Hg)
Melting Pt: 165…169 °C
Flash Pt: 265.8 °C
Storage Temperature: Ambient
MDL Number: MFCD04039875
CAS Number: 496786-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty