Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(6-((6-bromo-8-cyclopentyl-5-methyl-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidin-2-yl)amino)pyridin-3-yl)piperazine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A241688-1G A241688-5G A241688-5G A241688-250MG A241688-25G A241688-1G
103811-422EA 22 USD InStock
103811-422 103851-226 77090-392 77193-344 77193-346 77304-216
tert-Butyl 4-(6-((6-bromo-8-cyclopentyl-5-methyl-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidin-2-yl)amino)pyridin-3-yl)piperazine-1-carboxylate 97%
tert-Butyl 4-(6-((6-bromo-8-cyclopentyl-5-methyl-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidin-2-yl)amino)pyridin-3-yl)piperazine-1-carboxylate
Precaución: for Research Use Only


Formula: C₂₇H₃₄BrN₇O₃
Boiling Pt: 720 °C (760 mm Hg)
Flash Pt: 389.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834002
CAS Number: 571188-82-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty