Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(bromomethyl)piperidine-1-carboxylate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A238495-1G A238495-5G A238495-100G A238495-100MG A238495-10G A238495-250MG A238495-1G A238495-25G A238495-5G A238495-500G
103811-232EA 17 USD InStock
103811-232 103851-014 77013-694 77013-696 77013-698 77013-700 77090-132 77090-134 77090-136 77683-718
tert-Butyl-4-(bromomethyl)piperidine-1-carboxylate 97%, Ambeed.Inc
tert-Butyl-4-(bromomethyl)piperidine-1-carboxylate
Precaución: For research use only.


Formula: C₁₁H₂₀BrNO₂
Boiling Pt: 318.3 °C (760 mm Hg)
Flash Pt: 146.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04115538
CAS Number: 158407-04-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty