Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(chloromethyl)benzoate 95%

Supplier: AMBEED, INC
Danger

A104450-1G A104450-5G A104450-100G A104450-100MG A104450-250MG A104450-25G A104450-5G
103796-710EA 10 USD InStock
103796-710 103822-424 76780-710 76780-712 76780-714 76780-716 77283-208
tert-Butyl-4-(chloromethyl)benzoate 95%
tert-Butyl 4-(chloromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₂H₁₅ClO₂
Boiling Pt: 310 °C (760 mm Hg)
Flash Pt: 320 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15142807
CAS Number: 121579-86-0
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty