Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester 98%

Supplier: AMBEED, INC
Warning

A146297-1G A146297-5G A146297-100G A146297-100MG A146297-10G A146297-250MG A146297-1G A146297-25G A146297-5G
103803-652EA 18 USD InStock
103803-652 103841-738 76719-526 76719-528 76719-530 76719-532 77317-056 77317-058 77317-060
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester 98%
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₃O₂
Boiling Pt: 366 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08061257
CAS Number: 657428-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty