Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-amino-4-ethylpiperidine-1-carboxylate 96%

Supplier: AMBEED, INC
Warning

A572860-1G A572860-5G
103828-812EA 292 USD InStock
103828-812 103839-954 76844-082 76844-084
tert-Butyl 4-amino-4-ethylpiperidine-1-carboxylate 96%
tert-Butyl 4-amino-4-ethylpiperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₂₄N₂O₂
MW: 228.33 g/mol
Boiling Pt: 300.561 °C (760 mm Hg)
Flash Pt: 135.575 °C
Storage Temperature: Ambient
MDL Number: MFCD12408566
CAS Number: 741687-07-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty