Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-bromo-2,6-difluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A684468-100MG A684468-250MG A684468-25G A684468-1G A684468-5G
103840-550EA 66 USD InStock
103840-550 76790-986 76790-988 76790-990 76844-754 76844-756
tert-Butyl 4-bromo-2,6-difluorobenzoate 97%
tert-Butyl 4-bromo-2,6-difluorobenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₁BrF₂O₂
MW: 293.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08703422
CAS Number: 955887-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty