Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-bromobenzoate 97%

Supplier: AMBEED, INC
Warning

A117209-1G A117209-5G A117209-100G A117209-25G A117209-1G A117209-5G
103798-848EA 9 USD InStock
103798-848 103824-720 77009-238 77009-240 77311-508 77311-510
tert-Butyl-4-bromobenzoate 97%
tert-Butyl 4-bromobenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₂
MW: 257.1200 g/mol
Boiling Pt: 297 °C (760 mm Hg)
Flash Pt: 133.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00093266
CAS Number: 59247-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty