Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-bromoisoindoline-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A152606-1G A152606-5G A152606-100MG A152606-10G A152606-250MG A152606-1G A152606-25G A152606-5G
103804-408EA 47 USD InStock
103804-408 103842-580 76812-282 76812-284 76812-286 77318-198 77318-200 77318-202
tert-Butyl 4-bromoisoindoline-2-carboxylate 95%
tert-Butyl 4-bromoisoindoline-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₆BrNO₂
Boiling Pt: 363.2 °C (760 mm Hg)
Flash Pt: 173.469 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10700130
CAS Number: 1035235-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty