Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-chloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 95%

Supplier: AMBEED, INC
Warning

A117561-1G A117561-1G A117561-100MG A117561-250MG A117561-25G
103798-910EA 77 USD InStock
103798-910 103824-798 77311-580 77871-501 77871-502 77871-503
tert-Butyl-4-chloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 95%
tert-Butyl-4-chloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₆ClN₃O₂
MW: 269.7300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11109465
CAS Number: 1053656-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty