Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-chloropicolinate 98%

Supplier: AMBEED, INC
Warning

A111079-1G A111079-5G A111079-100MG A111079-250MG A111079-1G
103797-790EA 16 USD InStock
103797-790 103823-600 77284-084 77284-086 77310-408
tert-Butyl 4-chloropicolinate 98%
tert-Butyl 4-chloropicolinate
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClNO₂
MW: 213.66 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10565714
CAS Number: 220000-86-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty