Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-cyanobenzylcarbamate 97%

Supplier: AMBEED, INC
Warning

A289339-1G A289339-5G A289339-10G A289339-250MG
103813-826EA 20 USD InStock
103813-826 103834-966 76787-518 76787-520
tert-Butyl 4-cyanobenzylcarbamate 97%
tert-Butyl 4-cyanobenzylcarbamate
Precaución: for Research Use Only


Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
Boiling Pt: 392.9 °C (760 mm Hg)
Flash Pt: 191.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436197
CAS Number: 66389-80-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty