Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-ethynylpiperidine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A342456-1G A342456-100G A342456-10G A342456-250MG A342456-1G A342456-25G A342456-5G
103825-862EA 11 USD InStock
103825-862 103836-838 76788-290 76788-292 76788-294 76788-296 76839-876 76839-878 76839-880
tert-Butyl-4-ethynylpiperidine-1-carboxylate 97%
tert-Butyl-4-ethynylpiperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₉NO₂
Boiling Pt: 269.5 °C (760 mm Hg)
Flash Pt: 116.84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038026
CAS Number: 287192-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty