Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-hydroxybenzoate 96%

Supplier: AMBEED, INC
Warning

A432876-5G A432876-250MG A432876-1G A432876-100G
103827-538EA 6 USD InStock
103827-538 103838-604 76789-230 76842-304 77872-692
tert-Butyl 4-hydroxybenzoate 96%
tert-Butyl 4-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09953072
CAS Number: 25804-49-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty