Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-5-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%

Supplier: AMBEED, INC
Warning

A109239-1G A109239-5G A109239-250MG A109239-25G A109239-1G A109239-5G A109239-100G
103797-474EA 14 USD InStock
103797-474 103823-266 76765-686 76765-688 77310-092 77310-094 77876-821
tert-Butyl-5-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%
tert-Butyl-5-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₈BrNO₂
Flash Pt: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08752227
CAS Number: 215184-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty