Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-5-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%

Supplier: AMBEED, INC
Warning

A127124-1G A127124-100MG A127124-250MG A127124-1G
103800-914EA 117 USD InStock
103800-914 76782-092 76782-094 77298-176
tert-Butyl-5-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%
tert-Butyl-5-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate
Precaución: for Research Use Only


Formula: C₁₇H₂₁ClN₂O₃
Boiling Pt: 501.5±50.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10566053
CAS Number: 2103402-31-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty