Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A101025-1G A101025-5G A101025-100MG A101025-250MG
103795-760EA 64 USD InStock
103795-760 103821-452 77008-300 77008-302
tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate 97%
tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11045243
CAS Number: 957204-30-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty