Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Boc-6-oxo-2-azaspiro[3.4]octane 96%

Supplier: AMBEED, INC
Warning

A455281-5G A455281-1G A455281-250MG
103827-834EA 53 USD InStock
103827-834 103838-904 76842-750 77156-160
2-Boc-6-oxo-2-azaspiro[3.4]octane 96%
2-Boc-6-oxo-2-azaspiro[3.4]octane
Precaución: for Research Use Only


Formula: C₁₂H₁₉NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD22421762
CAS Number: 1363382-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty