Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-7-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%

Supplier: AMBEED, INC
Warning

A290501-1G A290501-100MG A290501-10G A290501-250MG A290501-25G A290501-1G A290501-5G
103813-868EA 23 USD InStock
103813-868 103835-010 76787-546 76787-548 76787-550 76787-552 76837-592 76837-594
tert-Butyl-7-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%
tert-Butyl-7-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₄H₂₀N₂O₂
Boiling Pt: 394.7 °C (760 mm Hg)
Flash Pt: 192.53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973401
CAS Number: 171049-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty