Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-7-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%

Supplier: AMBEED, INC
Warning

A256093-1G A256093-100G A256093-10G A256093-250MG A256093-1G A256093-25G A256093-5G
103812-240EA 24 USD InStock
103812-240 103852-102 77031-696 77031-698 77031-700 77091-576 77091-578 77091-580
tert-Butyl-7-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%
tert-Butyl-7-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₈BrNO₂
Boiling Pt: 374.2 °C (760 mm Hg)
Flash Pt: 180.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08752229
CAS Number: 258515-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty