Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tert-butyl 8-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate 98%

Supplier: AMBEED, INC
Warning

A135343-1G A135343-100MG A135343-250MG A135343-25G A135343-1G A135343-5G
103802-140EA 22 USD InStock
103802-140 76823-194 76823-196 76823-198 77314-956 77314-958
Tert-butyl 8-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate 98%
Tert-butyl 8-bromo-3,4-dihydroisoquinoline-2(1H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08752230
CAS Number: 893566-75-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty