Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl furan-3-ylcarbamate 95%

Supplier: AMBEED, INC
Warning

A325547-1G A325547-250MG A325547-1G A325547-5G A325547-10G
103825-406EA 26 USD InStock
103825-406 103836-352 76693-844 76839-292 76839-294 77626-606
tert-Butyl furan-3-ylcarbamate 95%
tert-Butyl furan-3-ylcarbamate
Precaución: for Research Use Only


Formula: C₉H₁₃NO₃
MW: 183.2044 g/mol
Melting Pt: 136.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09839085
CAS Number: 56267-48-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty