Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl methoxycarbamate 98%

Supplier: AMBEED, INC
Warning

A531705-1G A531705-5G A531705-250MG
103828-572EA 84 USD InStock
103828-572 103839-684 76843-700 76843-702 77169-600
tert-Butyl methoxycarbamate 98%
tert-Butyl methoxycarbamate
Precaución: for Research Use Only


Formula: C₆H₁₃NO₃
MW: 147.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21140572
CAS Number: 58377-44-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty