Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-N,N'-dimethylethylamine 97%

Supplier: AMBEED, INC
Danger

A129258-1G A129258-10G A129258-1G A129258-25G A129258-5G
103801-250EA 5 USD InStock
103801-250 76811-088 77313-628 77313-630 77313-632
N-Boc-N,N'-dimethylethylamine 97%
N-Boc-N,N'-dimethylethylamine
Precaución: for Research Use Only


Formula: C₉H₂₀N₂O₂
Boiling Pt: 244.5±19.0 °C (760 mm Hg)
Flash Pt: 101.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09953267
CAS Number: 112257-19-9
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty