Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl oxazol-5-ylcarbamate 97%

Supplier: AMBEED, INC
Warning

A135159-1G A135159-100MG A135159-250MG
103802-100EA 74 USD InStock
103802-100 76811-472 76811-474
tert-Butyl oxazol-5-ylcarbamate 97%
tert-Butyl oxazol-5-ylcarbamate
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O₃
Boiling Pt: 225.3±13.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD19688646
CAS Number: 1346809-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty