Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl piperazin-1-ylcarbamate 97%

Supplier: AMBEED, INC
Warning

A363235-1G A363235-5G A363235-100MG A363235-250MG A363235-10G
103826-338EA 80 USD InStock
103826-338 103837-358 76840-580 77033-210 77033-212 77872-554
tert-Butyl piperazin-1-ylcarbamate 97%
tert-Butyl piperazin-1-ylcarbamate
Precaución: for Research Use Only


Formula: C₉H₁₉N₃O₂
MW: 201.2661 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11840387
CAS Number: 147081-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty