Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl(2-iodoethoxy)dimethylsilane 97%

Supplier: AMBEED, INC
Warning

A108333-1G A108333-100G A108333-10G A108333-25G A108333-1G A108333-5G
103797-326EA 6 USD InStock
103797-326 103823-108 77025-586 77025-588 77025-590 77309-952 77309-954
tert-Butyl(2-iodoethoxy)dimethylsilane 97%
tert-Butyl(2-iodoethoxy)dimethylsilane
Precaución: for Research Use Only


Formula: C₈H₁₉IOSi
MW: 286.23 g/mol
Boiling Pt: 209.7 °C (760 mm Hg)
Flash Pt: 80.681 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909940
CAS Number: 101166-65-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty