Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl(3-chloropropoxy)dimethylsilane 97%

Supplier: AMBEED, INC
Warning

A220159-1G A220159-5G A220159-100G A220159-10G A220159-1G A220159-25G
103818-838EA 5 USD InStock
103818-838 103849-720 76987-020 76987-022 77088-312 77088-314
tert-Butyl(3-chloropropoxy)dimethylsilane 97%
tert-Butyl(3-chloropropoxy)dimethylsilane
Precaución: for Research Use Only


Formula: C₉H₂₁ClOSi
MW: 208.8 g/mol
Boiling Pt: 165 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09909983
CAS Number: 89031-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty