Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyldimethylsilane 95%

Supplier: AMBEED, INC
Danger

A761260-5G A761260-10G A761260-1G A761260-25G
103840-886EA 36 USD InStock
103840-886 76791-418 76845-118 77308-270
tert-Butyldimethylsilane 95%
tert-Butyldimethylsilane
Precaución: for Research Use Only


Formula: C₆H₁₆Si
MW: 116.28 g/mol
Boiling Pt: 81…83 °C (760 mm Hg)
Flash Pt: –10.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010754
CAS Number: 29681-57-0
UN: 1993
ADR: 3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty