Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylammonium chloride 97%

Supplier: AMBEED, INC
Warning

A181677-1000G A181677-100G A181677-500G A181677-25G
103809-158EA 69 USD InStock
103809-158 76784-598 76784-600 77301-740
Tetraethylammonium chloride 97%
Tetraethylammonium chloride
Precaución: for Research Use Only


Formula: C₈H₂₀ClN
Melting Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00011828
CAS Number: 56-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty