Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxytetrahydropyran 97%

Supplier: AMBEED, INC
Warning

A142100-1G A142100-100MG A142100-250MG A142100-25G A142100-1G A142100-5G
103803-100EA 15 USD InStock
103803-100 77010-386 77010-388 77010-390 77316-312 77316-314
3-Hydroxytetrahydropyran 97%
3-Hydroxytetrahydropyran
Precaución: for Research Use Only


Formula: C₅H₁₀O₂
Boiling Pt: 189.416 °C (760 mm Hg)
Flash Pt: 84.775 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00219387
CAS Number: 19752-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty