Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminotetrahydropyran hydrochloride 97%

Supplier: AMBEED, INC
Warning

A155330-1G A155330-5G A155330-10G A155330-1G A155330-5G A155330-25G
103804-744EA 11 USD InStock
103804-744 103842-938 77095-934 77300-110 77300-112 77318-656
4-Aminotetrahydropyran hydrochloride 97%
4-Aminotetrahydropyran hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClNO
Boiling Pt: 151.4 °C (760 mm Hg)
Melting Pt: 230…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00100881
CAS Number: 33024-60-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty