Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Tetrahydrofuran-3-carboxylic acid 97% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm

A131779-1G A131779-100G A131779-10G A131779-1G A131779-25G A131779-5G
103801-628EA 6 USD InStock
103801-628 77109-286 77109-288 77314-148 77314-150 77314-152
(±)-Tetrahydrofuran-3-carboxylic acid 97% stabilised
(±)-Tetrahydro-2-furoic acid
Precaución: for Research Use Only


Formula: C₅H₈O₃
Boiling Pt: 140 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075173
CAS Number: 89364-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty